Product Details

SNP ID
rs56326322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:77034347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTAAAACAATTATTTGTAACG[A/G]TAGGAGACTGGTATATATATTATTA
Phenotype
MIM: 608464
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGGF1 PubMed Links

Gene Details

Gene
AGGF1
Gene Name
angiogenic factor with G-patch and FHA domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018046.4 Intron NP_060516.2

View Full Product Details