Product Details

SNP ID
rs60969440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89327334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTTGCCAGAATATACATCCAGT[C/T]TTAGCTGTATGCTGGAATTAAGGAG
Phenotype
MIM: 616175
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2J1 PubMed Links

Gene Details

Gene
UBE2J1
Gene Name
ubiquitin conjugating enzyme E2 J1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016021.2 3633 UTR 3 NP_057105.2
XM_011535887.1 3633 UTR 3 XP_011534189.1
XM_011535888.2 3633 UTR 3 XP_011534190.1

View Full Product Details