Product Details

SNP ID
rs55898944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159682179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCAATAGAATAGGACTAGAAAG[A/G]CATCCCTACAAGTCCCCAAAGTATA
Phenotype
MIM: 147460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SOD2 PubMed Links

Gene Details

Gene
SOD2
Gene Name
superoxide dismutase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000636.3 964 UTR 3 NP_000627.2
NM_001024465.2 964 Intron NP_001019636.1
NM_001024466.2 964 Intron NP_001019637.1
NM_001322814.1 964 UTR 3 NP_001309743.1
NM_001322815.1 964 UTR 3 NP_001309744.1
NM_001322816.1 964 Intron NP_001309745.1
NM_001322817.1 964 Intron NP_001309746.1
NM_001322819.1 964 UTR 3 NP_001309748.1
NM_001322820.1 964 UTR 3 NP_001309749.1

View Full Product Details