Product Details

SNP ID
rs56113469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30949088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCAGCTCACATAAGACTACAAC[C/T]TCCTTCCACAACTCAGGCAATTCAC
Phenotype
MIM: 613928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPCR1 PubMed Links

Gene Details

Gene
DPCR1
Gene Name
diffuse panbronchiolitis critical region 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080870.3 624 Silent Mutation ACC,ACT T208T NP_543146.2

View Full Product Details