Product Details

SNP ID
rs58630663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:26644663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCAGACACTGTAAATTGAGTGC[G/T]TGAAGCTGTGAGAATAAACCCATCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C7orf71 PubMed Links

Gene Details

Gene
C7orf71
Gene Name
chromosome 7 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145531.2 Intron NP_001139003.1
NM_001302625.1 Intron NP_001289554.1
XM_017012010.1 Intron XP_016867499.1
XM_017012011.1 Intron XP_016867500.1
XM_017012012.1 Intron XP_016867501.1
XM_017012013.1 Intron XP_016867502.1
XM_017012014.1 Intron XP_016867503.1

View Full Product Details