Product Details

SNP ID
rs56410734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44565893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAAAGGGCCCAGCACTGCCGGCC[C/T]CAGAACTGTCTGTTCAGGCTGTGCA
Phenotype
MIM: 608023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX56 PubMed Links

Gene Details

Gene
DDX56
Gene Name
DEAD-box helicase 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257189.1 2276 UTR 3 NP_001244118.1
NM_019082.3 2276 UTR 3 NP_061955.1

View Full Product Details