Product Details
- SNP ID
-
rs62481482
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:152139714 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTCTCTTTCCTGTTGGCTACTTC[A/G]TTTCGAATGATAGTCCCGATGTACT
- Phenotype
-
MIM: 606833
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KMT2C
PubMed Links
Gene Details
- Gene
- KMT2C
- Gene Name
- lysine methyltransferase 2C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_170606.2 |
14835 |
Silent Mutation |
AAC,AAT |
N4807N |
NP_733751.2 |
XM_005250025.4 |
14835 |
Silent Mutation |
AAC,AAT |
N4879N |
XP_005250082.1 |
XM_005250026.3 |
14835 |
Silent Mutation |
AAC,AAT |
N4878N |
XP_005250083.1 |
XM_005250027.4 |
14835 |
Silent Mutation |
AAC,AAT |
N4878N |
XP_005250084.1 |
XM_005250028.4 |
14835 |
Silent Mutation |
AAC,AAT |
N4875N |
XP_005250085.1 |
XM_005250031.4 |
14835 |
Silent Mutation |
AAC,AAT |
N4824N |
XP_005250088.1 |
XM_006716077.3 |
14835 |
Silent Mutation |
AAC,AAT |
N4878N |
XP_006716140.1 |
XM_006716078.3 |
14835 |
Silent Mutation |
AAC,AAT |
N4855N |
XP_006716141.1 |
XM_006716079.3 |
14835 |
Silent Mutation |
AAC,AAT |
N4823N |
XP_006716142.1 |
XM_011516450.2 |
14835 |
Silent Mutation |
AAC,AAT |
N4863N |
XP_011514752.1 |
XM_011516451.2 |
14835 |
Silent Mutation |
AAC,AAT |
N4839N |
XP_011514753.1 |
XM_011516452.2 |
14835 |
Silent Mutation |
AAC,AAT |
N4828N |
XP_011514754.1 |
XM_011516453.2 |
14835 |
Silent Mutation |
AAC,AAT |
N4800N |
XP_011514755.1 |
XM_011516454.2 |
14835 |
Silent Mutation |
AAC,AAT |
N4574N |
XP_011514756.1 |
XM_011516456.2 |
14835 |
Silent Mutation |
AAC,AAT |
N4859N |
XP_011514758.1 |
XM_017012480.1 |
14835 |
Silent Mutation |
AAC,AAT |
N4879N |
XP_016867969.1 |
XM_017012481.1 |
14835 |
Silent Mutation |
AAC,AAT |
N4878N |
XP_016867970.1 |
XM_017012482.1 |
14835 |
Silent Mutation |
AAC,AAT |
N4874N |
XP_016867971.1 |
XM_017012483.1 |
14835 |
Silent Mutation |
AAC,AAT |
N4874N |
XP_016867972.1 |
XM_017012484.1 |
14835 |
Silent Mutation |
AAC,AAT |
N4868N |
XP_016867973.1 |
XM_017012485.1 |
14835 |
Silent Mutation |
AAC,AAT |
N4862N |
XP_016867974.1 |
XM_017012486.1 |
14835 |
Silent Mutation |
AAC,AAT |
N4850N |
XP_016867975.1 |
XM_017012487.1 |
14835 |
Silent Mutation |
AAC,AAT |
N4830N |
XP_016867976.1 |
XM_017012488.1 |
14835 |
Silent Mutation |
AAC,AAT |
N4818N |
XP_016867977.1 |
XM_017012489.1 |
14835 |
Silent Mutation |
AAC,AAT |
N3769N |
XP_016867978.1 |
XM_017012490.1 |
14835 |
Silent Mutation |
AAC,AAT |
N3637N |
XP_016867979.1 |
- Gene
- LOC731075
- Gene Name
- uncharacterized LOC731075
There are no transcripts associated with this gene.
View Full Product Details