Product Details

SNP ID
rs59531993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42837159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGGGAACTGGATGTCCTTCAG[C/T]TAGGGTGTAATGAAAGAAAAAAGCG
Phenotype
MIM: 609520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THAP1 PubMed Links

Gene Details

Gene
THAP1
Gene Name
THAP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018105.2 1682 UTR 3 NP_060575.1
NM_199003.1 1682 UTR 3 NP_945354.1

View Full Product Details