Product Details

SNP ID
rs59303802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:47777793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGTACCATGATGCTGTACATAA[G/T]GCCCGTTTCTTTCATTGGTAACATC
Phenotype
MIM: 600899
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRKDC PubMed Links

Gene Details

Gene
PRKDC
Gene Name
protein kinase, DNA-activated, catalytic polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081640.1 11899 Missense Mutation ATT,CTT I3948L NP_001075109.1
NM_006904.6 11899 Missense Mutation ATT,CTT I3979L NP_008835.5

View Full Product Details