Product Details

SNP ID
rs56090750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:47778503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGCCCAAAGTCGATCCCGATCA[C/T]GCCGCCAGTCTCCATGGCCACCATA
Phenotype
MIM: 600899
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRKDC PubMed Links

Gene Details

Gene
PRKDC
Gene Name
protein kinase, DNA-activated, catalytic polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081640.1 11773 Missense Mutation ATG,GTG M3906V NP_001075109.1
NM_006904.6 11773 Missense Mutation ATG,GTG M3937V NP_008835.5

View Full Product Details