Product Details

SNP ID
rs61051202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101534669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGATGATAACTGACTTGTCTAGT[A/G]CTAATGACTTTAGTAGGCATTTCAC
Phenotype
MIM: 607699
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF20 PubMed Links

Gene Details

Gene
RNF20
Gene Name
ring finger protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019592.6 Intron NP_062538.5
XM_011518862.1 Intron XP_011517164.1

View Full Product Details