Product Details

SNP ID
rs55985529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128822370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCCTACGTAAAGTACCAGGTCAT[C/T]GGCAAGAACCACGTGGCAGTGCCCA
Phenotype
MIM: 600440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENDOG PubMed Links

Gene Details

Gene
ENDOG
Gene Name
endonuclease G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004435.2 1567 Silent Mutation ATC,ATT I218I NP_004426.2
XM_011518347.2 1567 UTR 3 XP_011516649.1
Gene
SPOUT1
Gene Name
SPOUT domain containing methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016390.3 1567 UTR 3 NP_057474.2
XM_017014804.1 1567 UTR 3 XP_016870293.1

View Full Product Details