Product Details

SNP ID
rs57444011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71747689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGGGGTGGATCAAACAAGTTA[C/T]AACACATGTGAAAGTGTGTTGGAAA
Phenotype
MIM: 615608 MIM: 605516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf105 PubMed Links

Gene Details

Gene
C10orf105
Gene Name
chromosome 10 open reading frame 105
There are no transcripts associated with this gene.

Gene
C10orf54
Gene Name
chromosome 10 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022153.1 4641 UTR 3 NP_071436.1
Gene
CDH23
Gene Name
cadherin-related 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171930.1 4641 Intron NP_001165401.1
NM_001171931.1 4641 Intron NP_001165402.1
NM_001171932.1 4641 Intron NP_001165403.1
NM_001171933.1 4641 Intron NP_001165404.1
NM_001171934.1 4641 Intron NP_001165405.1
NM_001171935.1 4641 Intron NP_001165406.1
NM_001171936.1 4641 Intron NP_001165407.1
NM_022124.5 4641 Intron NP_071407.4
NM_052836.3 4641 Intron NP_443068.1
XM_006717940.3 4641 Intron XP_006718003.1
XM_006717942.3 4641 Intron XP_006718005.1
XM_011540039.2 4641 Intron XP_011538341.1
XM_011540042.2 4641 Intron XP_011538344.1
XM_011540043.2 4641 Intron XP_011538345.1
XM_011540044.2 4641 Intron XP_011538346.1
XM_011540045.2 4641 Intron XP_011538347.1
XM_011540046.2 4641 Intron XP_011538348.1
XM_011540047.2 4641 Intron XP_011538349.1
XM_011540048.2 4641 Intron XP_011538350.1
XM_011540049.2 4641 Intron XP_011538351.1
XM_011540051.2 4641 Intron XP_011538353.1
XM_011540052.2 4641 Intron XP_011538354.1
XM_017016499.1 4641 Intron XP_016871988.1
XM_017016500.1 4641 Intron XP_016871989.1
XM_017016501.1 4641 Intron XP_016871990.1
XM_017016502.1 4641 Intron XP_016871991.1
XM_017016503.1 4641 Intron XP_016871992.1
XM_017016504.1 4641 Intron XP_016871993.1
XM_017016505.1 4641 Intron XP_016871994.1
XM_017016506.1 4641 Intron XP_016871995.1
XM_017016507.1 4641 Intron XP_016871996.1
XM_017016508.1 4641 Intron XP_016871997.1

View Full Product Details