Product Details

SNP ID
rs55716129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:52765196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATAATTTTACACAAATTATTTTA[A/G]ATAATAAGGAGCATGAGAGCACATC
Phenotype
MIM: 154545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MBL2 PubMed Links

Gene Details

Gene
MBL2
Gene Name
mannose binding lectin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000242.2 3821 Intron NP_000233.1
XM_006717861.3 3821 UTR 3 XP_006717924.1
XM_011539816.2 3821 UTR 3 XP_011538118.1

View Full Product Details