Product Details

SNP ID
rs57562594
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:22208910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCAAATCCCGAAATCCCATAACC[A/G]CGAAGGATGTTAAGTGTATATGGAT
Phenotype
MIM: 613249
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
EBLN1 PubMed Links

Gene Details

Gene
EBLN1
Gene Name
endogenous Bornavirus-like nucleoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199938.1 1074 Silent Mutation CGC,CGT R358R NP_001186867.1

View Full Product Details