Product Details

SNP ID
rs61882399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17788670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTCCAAGCGTGGCTTTCCTCCT[C/T]GGCCCTTCCCTACTGTCACCACCAC
Phenotype
MIM: 176258 MIM: 606051
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNC1 PubMed Links

Gene Details

Gene
KCNC1
Gene Name
potassium voltage-gated channel subfamily C member 1
There are no transcripts associated with this gene.

Gene
SERGEF
Gene Name
secretion regulating guanine nucleotide exchange factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012139.3 Intron NP_036271.1

View Full Product Details