Product Details
- SNP ID
-
rs57867094
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:26667165 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATGCTAGATAAAAGCAGTTAACT[C/G]TATAGTAAACCTAGCTAATTTGCAA
- Phenotype
-
MIM: 610110
MIM: 612455
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ANO3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143895446,rs147516628] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANO3
- Gene Name
- anoctamin 3
- Gene
- SLC5A12
- Gene Name
- solute carrier family 5 member 12
View Full Product Details