Product Details

SNP ID
rs58353604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8612861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACTACAGCAGGCTCTAACTGGT[A/G]CAGGATCATGGTGAGGAGCTGCCCA
Phenotype
MIM: 612000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM66 PubMed Links

Gene Details

Gene
TRIM66
Gene Name
tripartite motif containing 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014818.1 9783 UTR 3 NP_055633.1
XM_006718397.2 9783 UTR 3 XP_006718460.1
XM_006718398.3 9783 UTR 3 XP_006718461.1
XM_011520504.1 9783 UTR 3 XP_011518806.1
XM_011520507.2 9783 UTR 3 XP_011518809.1
XM_011520508.1 9783 UTR 3 XP_011518810.1
XM_011520509.1 9783 UTR 3 XP_011518811.1
XM_011520510.1 9783 UTR 3 XP_011518812.1
XM_011520511.1 9783 UTR 3 XP_011518813.1
XM_011520512.1 9783 UTR 3 XP_011518814.1
XM_011520513.1 9783 UTR 3 XP_011518815.1
XM_011520514.2 9783 UTR 3 XP_011518816.1
XM_011520515.1 9783 UTR 3 XP_011518817.1
XM_011520516.1 9783 UTR 3 XP_011518818.1
XM_011520517.2 9783 UTR 3 XP_011518819.1
XM_011520518.1 9783 UTR 3 XP_011518820.1
XM_011520519.1 9783 UTR 3 XP_011518821.1
XM_011520522.2 9783 UTR 3 XP_011518824.1
XM_011520523.2 9783 UTR 3 XP_011518825.1
XM_011520524.1 9783 Intron XP_011518826.1
XM_011520525.1 9783 Intron XP_011518827.1
XM_011520526.2 9783 UTR 3 XP_011518828.1
XM_011520527.1 9783 UTR 3 XP_011518829.1
XM_017018629.1 9783 UTR 3 XP_016874118.1
XM_017018630.1 9783 UTR 3 XP_016874119.1
XM_017018631.1 9783 UTR 3 XP_016874120.1

View Full Product Details