Product Details

SNP ID
rs61108232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120712203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCAGCTTCTTAGAGTCTGGCA[A/G]CCTCTTTGTTACTCTTCTTAACGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UNC119B PubMed Links

Gene Details

Gene
UNC119B
Gene Name
unc-119 lipid binding chaperone B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080533.2 Intron NP_001074002.1

View Full Product Details