Product Details

SNP ID
rs61962247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:94441065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGCCAACTGTTCTTAAATTCAG[C/T]TCCTCTGAATTTGTTCTATACCTTA
Phenotype
MIM: 191275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCT PubMed Links

Gene Details

Gene
DCT
Gene Name
dopachrome tautomerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129889.2 Intron NP_001123361.1
NM_001322182.1 Intron NP_001309111.1
NM_001322183.1 Intron NP_001309112.1
NM_001322184.1 Intron NP_001309113.1
NM_001322185.1 Intron NP_001309114.1
NM_001322186.1 Intron NP_001309115.1
NM_001922.4 Intron NP_001913.2
XM_011521049.1 Intron XP_011519351.1
XM_017020401.1 Intron XP_016875890.1

View Full Product Details