Product Details

SNP ID
rs62020698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42945216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATTGATGTAAGTGAACCTGGA[C/T]ATGGAGCAAAAGACCCTCCAATACT
Phenotype
MIM: 605981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBR1 PubMed Links

Gene Details

Gene
UBR1
Gene Name
ubiquitin protein ligase E3 component n-recognin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174916.2 5429 UTR 3 NP_777576.1

View Full Product Details