Product Details
- SNP ID
-
rs61268760
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:43134398 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGGCTTGCCGTGCTGTGGTGGTG[G/T]TCTGTGGTCCGCATGGTAGAACTCT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM62
PubMed Links
Gene Details
- Gene
- TMEM62
- Gene Name
- transmembrane protein 62
View Full Product Details