Product Details

SNP ID
rs56835863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65327364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGGACTACCCTTCCCCAAGTC[A/C]ACGCGGACACAGCTCTAGTTTTCTA
Phenotype
MIM: 604184
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGDCC3 PubMed Links

Gene Details

Gene
IGDCC3
Gene Name
immunoglobulin superfamily DCC subclass member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004884.3 4181 UTR 3 NP_004875.2
XM_011522241.1 4181 UTR 3 XP_011520543.1
XM_011522243.1 4181 UTR 3 XP_011520545.1
XM_011522244.1 4181 UTR 3 XP_011520546.1
XM_011522245.2 4181 Intron XP_011520547.1

View Full Product Details