Product Details

SNP ID
rs62033352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20859890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTCCTGGGACAGAGCCACTAA[A/G]TCTGCTCCTCGGGACACAAGCCCTC
Phenotype
MIM: 616167
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCUN1D3 PubMed Links

Gene Details

Gene
DCUN1D3
Gene Name
defective in cullin neddylation 1 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173475.3 1171 Missense Mutation ACT,ATT T304I NP_775746.1

View Full Product Details