Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_005886.2 | 3918 | Intron | NP_005877.2 | ||
XM_005255772.4 | 3918 | Intron | XP_005255829.1 | ||
XM_006721121.3 | 3918 | Intron | XP_006721184.1 | ||
XM_006721122.2 | 3918 | Intron | XP_006721185.1 | ||
XM_006721123.3 | 3918 | Intron | XP_006721186.1 | ||
XM_011522810.2 | 3918 | Intron | XP_011521112.1 | ||
XM_017022860.1 | 3918 | Intron | XP_016878349.1 | ||
XM_017022861.1 | 3918 | Intron | XP_016878350.1 | ||
XM_017022862.1 | 3918 | Intron | XP_016878351.1 | ||
XM_017022863.1 | 3918 | Intron | XP_016878352.1 | ||
XM_017022864.1 | 3918 | Intron | XP_016878353.1 |