Product Details

SNP ID
rs58631882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75538665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAACAAGATTAATAAGTAAGCTA[G/T]TCAGTGTGTCCAAACGTGGCCAGGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LLGL2 PubMed Links

Gene Details

Gene
LLGL2
Gene Name
LLGL2, scribble cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015002.1 Intron NP_001015002.1
NM_001031803.1 Intron NP_001026973.1
NM_004524.2 Intron NP_004515.2
XM_011524801.1 Intron XP_011523103.1
XM_011524802.1 Intron XP_011523104.1
XM_011524807.2 Intron XP_011523109.1
XM_017024625.1 Intron XP_016880114.1
XM_017024626.1 Intron XP_016880115.1
XM_017024627.1 Intron XP_016880116.1
XM_017024628.1 Intron XP_016880117.1
XM_017024629.1 Intron XP_016880118.1
XM_017024630.1 Intron XP_016880119.1
XM_017024631.1 Intron XP_016880120.1
XM_017024632.1 Intron XP_016880121.1

View Full Product Details