Product Details

SNP ID
rs58556339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7922835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGACGAAGTGGCAGAGAGCCGAC[A/G]GCGAGTAGCTCATGCCCGGGATTTG
Phenotype
MIM: 611425 MIM: 604111 MIM: 610969
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNTROB PubMed Links

Gene Details

Gene
CNTROB
Gene Name
centrobin, centriole duplication and spindle assembly protein
There are no transcripts associated with this gene.

Gene
KCNAB3
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004732.3 1340 UTR 3 NP_004723.2
XM_011524068.1 1340 UTR 3 XP_011522370.1
XM_017025304.1 1340 UTR 3 XP_016880793.1
XM_017025305.1 1340 UTR 3 XP_016880794.1
XM_017025306.1 1340 UTR 3 XP_016880795.1
Gene
LOC284023
Gene Name
uncharacterized LOC284023
There are no transcripts associated with this gene.

Gene
TRAPPC1
Gene Name
trafficking protein particle complex 1
There are no transcripts associated with this gene.

View Full Product Details