Product Details

SNP ID
rs56929273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41515688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGCCAGTGAGTTCTCCAGCCCA[G/T]CTTTCTGGGGGAGTGACAGATGCAT
Phenotype
MIM: 148030 MIM: 148020
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRT15 PubMed Links

Gene Details

Gene
KRT15
Gene Name
keratin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002275.3 1289 Missense Mutation GAT,GCT D344A NP_002266.2
XM_011524784.2 1289 Missense Mutation GAT,GCT D351A XP_011523086.1
XM_017024614.1 1289 Missense Mutation GAT,GCT D351A XP_016880103.1
Gene
KRT19
Gene Name
keratin 19
There are no transcripts associated with this gene.

Gene
MIR6510
Gene Name
microRNA 6510
There are no transcripts associated with this gene.

View Full Product Details