Product Details

SNP ID
rs56851766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:55751432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGCGGTGCCTCCAAGTGACTGC[C/T]GTGCAGATCCAGGGGAGTTGGAAGG
Phenotype
MIM: 606055
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCTP PubMed Links

Gene Details

Gene
PCTP
Gene Name
phosphatidylcholine transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102402.2 303 UTR 5 NP_001095872.1
NM_021213.3 303 Intron NP_067036.2
XM_005257549.1 303 Intron XP_005257606.1
XM_005257550.3 303 Intron XP_005257607.1
XM_017024913.1 303 UTR 5 XP_016880402.1

View Full Product Details