Product Details

SNP ID
rs62635764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44908143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGACACAGACTTGGTGTCCAGG[A/C]TGGTTTCTGCAGATGTGGGGAGAGG
Phenotype
MIM: 614677 MIM: 603892 MIM: 137780
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC103 PubMed Links

Gene Details

Gene
CCDC103
Gene Name
coiled-coil domain containing 103
There are no transcripts associated with this gene.

Gene
EFTUD2
Gene Name
elongation factor Tu GTP binding domain containing 2
There are no transcripts associated with this gene.

Gene
GFAP
Gene Name
glial fibrillary acidic protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131019.2 1315 Intron NP_001124491.1
NM_001242376.1 1315 Intron NP_001229305.1
NM_002055.4 1315 Missense Mutation AGC,ATC S393I NP_002046.1
XM_017024451.1 1315 Missense Mutation AGC,ATC S433I XP_016879940.1

View Full Product Details