Product Details

SNP ID
rs61751982
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64480378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTTCTAGTAACTCATTAAATAG[A/C]CCTTGACAAACCTAGAAAGAAATAG
Phenotype
MIM: 604983
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
MILR1 PubMed Links

Gene Details

Gene
MILR1
Gene Name
mast cell immunoglobulin like receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085423.1 1301 Intron NP_001078892.1
NM_001291316.1 1301 Intron NP_001278245.1
NM_001291317.1 1301 Intron NP_001278246.1
Gene
POLG2
Gene Name
DNA polymerase gamma 2, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007215.3 1301 Silent Mutation GGG,GGT G401G NP_009146.2
XM_006721651.2 1301 Intron XP_006721714.1
XM_017024072.1 1301 Intron XP_016879561.1

View Full Product Details