Product Details

SNP ID
rs58471799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43528676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAGAGCTGGACGCTGATTGTCC[A/G]GGAAGGCCAAAGAGAAGAGGGCCTC
Phenotype
MIM: 600396 MIM: 600711
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHX8 PubMed Links

Gene Details

Gene
DHX8
Gene Name
DEAH-box helicase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302623.1 741 Intron NP_001289552.1
NM_001322216.1 741 Intron NP_001309145.1
NM_001322217.1 741 Intron NP_001309146.1
NM_001322218.1 741 Intron NP_001309147.1
NM_001322219.1 741 Intron NP_001309148.1
NM_001322220.1 741 Intron NP_001309149.1
NM_001322221.1 741 Intron NP_001309150.1
NM_004941.2 741 Intron NP_004932.1
Gene
ETV4
Gene Name
ETS variant 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079675.2 741 Missense Mutation CCG,CTG P433L NP_001073143.1
NM_001261437.1 741 Missense Mutation CCG,CTG P394L NP_001248366.1
NM_001261438.1 741 Missense Mutation CCG,CTG P394L NP_001248367.1
NM_001261439.1 741 Missense Mutation CCG,CTG P156L NP_001248368.1
NM_001986.2 741 Missense Mutation CCG,CTG P433L NP_001977.1
XM_011524514.1 741 Missense Mutation CCG,CTG P170L XP_011522816.1
XM_017024349.1 741 Missense Mutation CCG,CTG P428L XP_016879838.1

View Full Product Details