Product Details

SNP ID
rs59203084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32931585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGCCACCATCTTTCTGGCTTC[G/T]TTTGCAGCCTTGGTGCTGGTTTGCA
Phenotype
MIM: 615949
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM98 PubMed Links

Gene Details

Gene
TMEM98
Gene Name
transmembrane protein 98
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033504.1 439 Silent Mutation TCG,TCT S19S NP_001028676.1
NM_001301746.1 439 Silent Mutation TCG,TCT S19S NP_001288675.1
NM_015544.2 439 Silent Mutation TCG,TCT S19S NP_056359.2

View Full Product Details