Product Details

SNP ID
hCV89423233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39709262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCCCGCCAGCATCTGTGAAGAT[C/G]CTGCAAGTCTTCAGAGATATCTCCC
Phenotype
MIM: 607260
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LGALS14 PubMed Links

Gene Details

Gene
LGALS14
Gene Name
galectin 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020129.2 592 Missense Mutation ATC,ATG I123M NP_064514.1
NM_203471.1 592 Missense Mutation ATC,ATG I152M NP_982297.1

View Full Product Details