Product Details

SNP ID
rs56159111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43718986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTCTTCCTGCTGGTCTCCCCCC[G/T]CCGCTGCGGGGCCGTCGAGACCCGC
Phenotype
MIM: 613176
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IRGC PubMed Links

Gene Details

Gene
IRGC
Gene Name
immunity related GTPase cinema
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019612.3 609 Missense Mutation CGC,CTC R143L NP_062558.1
XM_005259058.3 609 Missense Mutation CGC,CTC R180L XP_005259115.1
XM_006723283.3 609 Missense Mutation CGC,CTC R143L XP_006723346.1
XM_011527118.2 609 Missense Mutation CGC,CTC R143L XP_011525420.1
Gene
SMG9
Gene Name
SMG9, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details