Product Details

SNP ID
rs60302252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50771062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTCTGGGCCCTGAACCACCGCAC[C/T]GTGCAGCGGGCGAGCTGGCTGGCCT
Phenotype
MIM: 603195
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR32 PubMed Links

Gene Details

Gene
GPR32
Gene Name
G protein-coupled receptor 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001506.2 599 Silent Mutation ACC,ACT T154T NP_001497.1
Gene
LOC105372440
Gene Name
uncharacterized LOC105372440
There are no transcripts associated with this gene.

View Full Product Details