Product Details

SNP ID
rs61505502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6773316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCTACTGTGGTTCATGCCCCCA[A/G]ATGGGACCAACCAGAGGTCTCGGGG
Phenotype
MIM: 604721 MIM: 164875
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SH2D3A PubMed Links

Gene Details

Gene
SH2D3A
Gene Name
SH2 domain containing 3A
There are no transcripts associated with this gene.

Gene
VAV1
Gene Name
vav guanine nucleotide exchange factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258206.1 Intron NP_001245135.1
NM_001258207.1 Intron NP_001245136.1
NM_005428.3 Intron NP_005419.2
XM_005259642.1 Intron XP_005259699.1

View Full Product Details