Product Details

SNP ID
rs62204276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:57608924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCATTGGTGGGACTGGGTGTGAT[C/T]CCTTATTCCAGATCCCAAAAGGGTA
Phenotype
MIM: 606750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBP1 PubMed Links

Gene Details

Gene
ZBP1
Gene Name
Z-DNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160417.1 Intron NP_001153889.1
NM_001160418.1 Intron NP_001153890.1
NM_001160419.2 Intron NP_001153891.1
NM_001323966.1 Intron NP_001310895.1
NM_030776.2 Intron NP_110403.2
XM_011529057.2 Intron XP_011527359.1
XM_011529058.2 Intron XP_011527360.1
XM_017028086.1 Intron XP_016883575.1

View Full Product Details