Product Details

SNP ID
rs56397561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:4220991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATAAATACAGCAGGCTTGTATGA[C/T]GGCTATACAATCTGCCACCAAAGCT
Phenotype
MIM: 104219
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADRA1D PubMed Links

Gene Details

Gene
ADRA1D
Gene Name
adrenoceptor alpha 1D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000678.3 2306 UTR 3 NP_000669.1

View Full Product Details