Product Details

SNP ID
rs56161101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29078468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGAGGCTGGAATTCAGAGTCTC[G/T]CTGGACTACAGAGAACGTGGCTTAG
Phenotype
MIM: 611110
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCT8 PubMed Links

Gene Details

Gene
CCT8
Gene Name
chaperonin containing TCP1 subunit 8
There are no transcripts associated with this gene.

Gene
MAP3K7CL
Gene Name
MAP3K7 C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286617.1 Intron NP_001273546.1
NM_001286618.1 Intron NP_001273547.1
NM_001286619.1 Intron NP_001273548.1
NM_001286620.1 Intron NP_001273549.1
NM_001286622.1 Intron NP_001273551.1
NM_001286623.1 Intron NP_001273552.1
NM_001286624.1 Intron NP_001273553.1
NM_001286634.1 Intron NP_001273563.1
NM_020152.3 Intron NP_064537.1

View Full Product Details