Product Details

SNP ID
rs58980844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:35038070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTGTACTTATTGACTTCCTCTG[C/T]GTGTCCATTGCTTTATACATATTGT
Phenotype
MIM: 151385
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RUNX1 PubMed Links

Gene Details

Gene
RUNX1
Gene Name
runt related transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001890.2 Intron NP_001001890.1
NM_001122607.1 Intron NP_001116079.1
NM_001754.4 Intron NP_001745.2
XM_005261068.3 Intron XP_005261125.1
XM_005261069.4 Intron XP_005261126.1
XM_011529766.2 Intron XP_011528068.1
XM_011529767.2 Intron XP_011528069.1
XM_011529768.2 Intron XP_011528070.1
XM_011529770.2 Intron XP_011528072.1
XM_017028487.1 Intron XP_016883976.1
Gene
RUNX1-IT1
Gene Name
RUNX1 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details