Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000489.4 | Intron | NP_000480.3 | |||
NM_138270.3 | Intron | NP_612114.2 | |||
XM_005262153.4 | Intron | XP_005262210.2 | |||
XM_005262154.4 | Intron | XP_005262211.2 | |||
XM_005262155.4 | Intron | XP_005262212.2 | |||
XM_005262156.4 | Intron | XP_005262213.2 | |||
XM_005262157.4 | Intron | XP_005262214.2 | |||
XM_006724666.3 | Intron | XP_006724729.1 | |||
XM_006724667.3 | Intron | XP_006724730.1 | |||
XM_006724668.2 | Intron | XP_006724731.1 | |||
XM_017029601.1 | Intron | XP_016885090.1 | |||
XM_017029602.1 | Intron | XP_016885091.1 | |||
XM_017029603.1 | Intron | XP_016885092.1 | |||
XM_017029604.1 | Intron | XP_016885093.1 | |||
XM_017029605.1 | Intron | XP_016885094.1 | |||
XM_017029606.1 | Intron | XP_016885095.1 | |||
XM_017029607.1 | Intron | XP_016885096.1 | |||
XM_017029608.1 | Intron | XP_016885097.1 | |||
XM_017029609.1 | Intron | XP_016885098.1 | |||
XM_017029610.1 | Intron | XP_016885099.1 | |||
XM_017029611.1 | Intron | XP_016885100.1 |