Product Details

SNP ID
rs61755732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70146758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCACCTTCAGAGGTGGATTGATA[C/T]CAGCTTAGAAGAGATTGAGAGCATT
Phenotype
MIM: 300139
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGBP1 PubMed Links

Gene Details

Gene
IGBP1
Gene Name
immunoglobulin (CD79A) binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001551.2 906 Missense Mutation ACC,ATC T203I NP_001542.1
XM_017029489.1 906 Missense Mutation ACC,ATC T203I XP_016884978.1
XM_017029490.1 906 Intron XP_016884979.1

View Full Product Details