Product Details
- SNP ID
-
rs62327420
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:141650410 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATAGTCAATACATATGGAGGTGCA[A/G]GCCCCAAACATACTGTTATCATGGA
- Phenotype
-
MIM: 600554
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL15
PubMed Links
Gene Details
- Gene
- IL15
- Gene Name
- interleukin 15
View Full Product Details