Product Details

SNP ID
hCV89709212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:144114690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACTGCAAATATTAACACCAACC[G/T]GGATTTTCTATTTCAACAGAACTTA
Phenotype
MIM: 111300
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GYPA PubMed Links

Gene Details

Gene
GYPA
Gene Name
glycophorin A (MNS blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308187.1 547 Silent Mutation CCA,CCC P132P NP_001295116.1
NM_001308190.1 547 Silent Mutation CCA,CCC P112P NP_001295119.1
NM_002099.7 547 Silent Mutation CCA,CCC P145P NP_002090.4

View Full Product Details