Product Details

SNP ID
rs56809525
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:118883843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCATAACCTGTAGTGGGGCGGGC[A/G]GGGTTTGGGGGGACAAACCAAAAAC
Phenotype
MIM: 604127
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBX15 PubMed Links

Gene Details

Gene
TBX15
Gene Name
T-box 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152380.2 3151 UTR 3 NP_689593.2
XM_005271161.3 3151 UTR 3 XP_005271218.1
XM_005271162.1 3151 UTR 3 XP_005271219.1
XM_011542052.2 3151 UTR 3 XP_011540354.1

View Full Product Details