Product Details

SNP ID
rs56292601
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:111308611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGAATTTCATTTTGTGACCAGTC[C/T]TTGTGAAAATGTAAATCCAAGTGCC
Phenotype
MIM: 606080
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHIA PubMed Links

Gene Details

Gene
CHIA
Gene Name
chitinase, acidic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040623.2 Intron NP_001035713.1
NM_001258001.1 Intron NP_001244930.1
NM_001258002.1 Intron NP_001244931.1
NM_001258003.1 Intron NP_001244932.1
NM_001258004.1 Intron NP_001244933.1
NM_001258005.1 Intron NP_001244934.1
NM_021797.3 Intron NP_068569.2
NM_201653.3 Intron NP_970615.2
XM_006710577.3 Intron XP_006710640.1
XM_017001047.1 Intron XP_016856536.1
XM_017001048.1 Intron XP_016856537.1

View Full Product Details