Product Details

SNP ID
rs61755355
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:111756683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTAAATCTGGCACCGGGAAAAC[C/T]TGTGTGTTCTCCACCATAGCTTTGG
Phenotype
MIM: 606168
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX20 PubMed Links

Gene Details

Gene
DDX20
Gene Name
DEAD-box helicase 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007204.4 696 Silent Mutation ACC,ACT T113T NP_009135.4
Gene
FAM212B
Gene Name
family with sequence similarity 212 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019099.4 696 Intron NP_061972.1
NM_198926.2 696 Intron NP_945120.1
XM_011541783.2 696 Intron XP_011540085.1
Gene
FAM212B-AS1
Gene Name
FAM212B antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC101928718
Gene Name
uncharacterized LOC101928718
There are no transcripts associated with this gene.

View Full Product Details