Product Details

SNP ID
rs59092658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:119765084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAATCTAGCGTCAAAAGCCTTGGG[C/T]AAAGTGTCTCATTTCTGAGTCAGGT
Phenotype
MIM: 176878
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPN4 PubMed Links

Gene Details

Gene
PTPN4
Gene Name
protein tyrosine phosphatase, non-receptor type 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002830.3 Intron NP_002821.1
XM_011511560.2 Intron XP_011509862.1
XM_011511562.2 Intron XP_011509864.1
XM_011511563.2 Intron XP_011509865.1
XM_017004596.1 Intron XP_016860085.1
XM_017004597.1 Intron XP_016860086.1
XM_017004598.1 Intron XP_016860087.1
XM_017004599.1 Intron XP_016860088.1
XM_017004600.1 Intron XP_016860089.1
XM_017004601.1 Intron XP_016860090.1
XM_017004602.1 Intron XP_016860091.1
XM_017004603.1 Intron XP_016860092.1
XM_017004604.1 Intron XP_016860093.1
XM_017004605.1 Intron XP_016860094.1
XM_017004606.1 Intron XP_016860095.1
XM_017004607.1 Intron XP_016860096.1
XM_017004608.1 Intron XP_016860097.1

View Full Product Details