Product Details
- SNP ID
-
hCV89908219
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:42762949 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTCCTTTTCCAGAGAGACCTCGC[C/G]CTGCACTTTCAGCTTCCCTATGGCC
- Phenotype
-
MIM: 604521
MIM: 609050
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HAAO
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79735204] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HAAO
- Gene Name
- 3-hydroxyanthranilate 3,4-dioxygenase
There are no transcripts associated with this gene.
- Gene
- MTA3
- Gene Name
- metastasis associated 1 family member 3
- Gene
- OXER1
- Gene Name
- oxoeicosanoid receptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_148962.4 |
1313 |
Missense Mutation |
CGC,GGC |
R411G |
NP_683765.1 |
View Full Product Details